Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HES-1 Antibody (HL2198) - Azide and BSA Free, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP325501
Description
HES-1 Monoclonal antibody specifically detects HES-1 in Human samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
HES-1 | |
Monoclonal | |
Unconjugated | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunocytochemistry | |
HL2198 | |
Western Blot 1:1000-1:10000, Immunocytochemistry/ Immunofluorescence | |
BHLHB39, bHLHb39hairy homolog (Drosophila), Class B basic helix-loop-helix protein 39, FLJ20408, Hairy and enhancer of split 1, hairy and enhancer of split 1, (Drosophila), Hairy homolog, Hairy-like protein, Hes1, HES-1, hHL, HL, HRYHHL, transcription factor HES-1 | |
Synthetic peptide of human HES-1. | |
100 μL | |
Stem Cell Markers | |
3280 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction