Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HES-1 Antibody (OTI4H1), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270937H
Description
HES-1 Monoclonal specifically detects HES-1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
HES-1 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
HES1 | |
Recombinant protein expressed in E.coli corresponding to amino acids 1-280 of human HES-1. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI4H1 | |
Western Blot, Immunohistochemistry | |
BHLHB39, bHLHb39hairy homolog (Drosophila), Class B basic helix-loop-helix protein 39, FLJ20408, Hairy and enhancer of split 1, hairy and enhancer of split 1, (Drosophila), Hairy homolog, Hairy-like protein, Hes1, HES-1, hHL, HL, HRYHHL, transcription factor HES-1 | |
Mouse | |
Affinity Purified | |
Neuronal Stem Cell Markers, Stem Cell Markers | |
3280.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction