Learn More
HES-1 Rat anti-Human,Mouse, Unconjugated, Clone: HS395A, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | HES-1 |
| Applications | ELISA, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | HS395A |
| Conjugate | Unconjugated |
| Gene Accession No. | NP_032261.1 |
| Gene Alias | Transcription factor HES-1;Hairy and enhancer of split 1 |
| Gene Symbols | Hes1, Hes-1 |
| Host Species | Rat |
| Immunogen | His-tagged recombinant fragment corresponding to 190 amino acids from murine HES-1 protein. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.