Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HES-1 Rat anti-Human,Mouse, Unconjugated, Clone: HS395A, MilliporeSigma™

Catalog No. MABN23182MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN23182MI 25 μL
MABN23181MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABN23182MI Supplier MilliporeSigma Supplier No. MABN231825UL
Add to Cart
Add to Cart

Rat Monoclonal Antibody

HES-1 Monoclonal antibody specifically detects HES-1 in Human,Mouse samples. It is validated for ELISA, Immunohistochemistry (Paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen HES-1
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone HS395A
Conjugate Unconjugated
Gene Accession No. NP_032261.1
Gene Alias Transcription factor HES-1;Hairy and enhancer of split 1
Gene Symbols Hes1, Hes-1
Host Species Rat
Immunogen His-tagged recombinant fragment corresponding to 190 amino acids from murine HES-1 protein.
Molecular Weight of Antigen 29.75 kDa calculated.
Purification Method Protein G Purified
Quantity 25 μL
Research Discipline Neuroscience
Primary or Secondary Primary
Test Specificity Clone HS395A is a rat monoclonal antibody that detects HES-1 in human and murine tissues.
Target Species Human, Mouse
Content And Storage Stable for 1 year at 2 to 8°C from date of receipt.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.