Learn More
HES1 Mouse anti-Human, Clone: OTI1B5, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF504046

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This protein belongs to the basic helix-loop-helix family of transcription factors. It is a transcriptional repressor of genes that require a bHLH protein for their transcription. The protein has a particular type of basic domain that contains a helix interrupting protein that binds to the N-box rather than the canonical E-box.Specifications
HES1 | |
Monoclonal | |
Unconjugated | |
HES1 | |
HES-1, HHL, HRY, bHLHb39 | |
Mouse | |
Affinity Chromatography | |
RUO | |
3280 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI1B5 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q14469 | |
HES1 | |
Full length human recombit protein of human HES1 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.