Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HES1 Mouse anti-Human, DyLight™ 488, Clone: OTI4H1, liquid, TrueMAB™
SDP

Catalog No. 501671298
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-1298 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-167-1298 Supplier OriGene Supplier No. TA400011
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This protein belongs to the basic helix-loop-helix family of transcription factors. It is a transcriptional repressor of genes that require a bHLH protein for their transcription. The protein has a particular type of basic domain that contains a helix interrupting protein that binds to the N-box rather than the canonical E-box.

Specifications

Antigen HES1
Applications Flow Cytometry
Classification Monoclonal
Clone OTI4H1
Concentration 0.52 mg/mL
Conjugate DyLight 488
Formulation PBS with 0.1% BSA, 50% glycerol and 0.02% sodium azide
Gene HES1
Gene Accession No. Q14469
Gene Alias bHLHb39, HES-1, HHL, HRY
Gene Symbols HES1
Host Species Mouse
Immunogen Recombit protein expressed in E.coli corresponding to amino acids 1-280 of human HES1
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3280
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.