Learn More
Invitrogen™ HES1 Polyclonal Antibody

Description
Immunogen sequence: PPLVPIPGGA APPPGGAPCK LGSQAGEAAK VFGGFQVVPA PDGQFAFLIP NGAFAHSGPV IPVYTSNSG Highest antigen sequence indentity to the following orthologs: Mouse - 99%, Rat - 97%.
Specifications
Specifications
| Antigen | HES1 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | HES1 |
| Gene Accession No. | Q14469 |
| Gene Alias | BHLHB39; class B basic helix-loop-helix protein 39; FLJ20408; Hairy and enhancer of split 1; hairy and enhancer of split 1 (Drosophila); hairy and enhancer of split 1,; hairy homolog; Hairy-like; Hairy-like protein; hes family bHLH transcription factor 1; HES1; Hes-1; hHL; HL; HRY; RHL; transcription factor HES-1 |
| Gene Symbols | HES1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.