Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HES1 Recombinant Rabbit Monoclonal Antibody (HL2198)
GREENER_CHOICE

Catalog No. FEMA550429
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550429 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550429 Supplier Invitrogen™ Supplier No. MA550429
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

HES1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF

This protein belongs to the basic helix-loop-helix family of transcription factors. It is a transcriptional repressor of genes that require a bHLH protein for their transcription. The protein has a particular type of basic domain that contains a helix interrupting protein that binds to the N-box rather than the canonical E-box.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HES1
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone HL2198
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene HES1
Gene Accession No. Q14469
Gene Alias BHLHB39; class B basic helix-loop-helix protein 39; FLJ20408; Hairy and enhancer of split 1; hairy and enhancer of split 1 (Drosophila); hairy and enhancer of split 1,; hairy homolog; Hairy-like; Hairy-like protein; hes family bHLH transcription factor 1; HES1; Hes-1; hHL; HL; HRY; RHL; transcription factor HES-1
Gene Symbols HES1
Host Species Rabbit
Immunogen Synthetic peptide of human HES1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3280
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.