Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HES2 Antibody, Janelia Fluor™ 549, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP227110JF549
Description
HES2 Polyclonal specifically detects HES2 in Human, Mouse, Rat, Primate samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
HES2 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
BHLHB40, bHLHb40Class B basic helix-loop-helix protein 40, Hairy and enhancer of split 2, hairy and enhancer of split 2 (Drosophila), transcription factor HES-2 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Store at 4°C in the dark. |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Janelia Fluor 549 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
HES2 | |
8 GDAAELRkSLkPLLEkRR 25 | |
0.1 mL | |
DNA replication Transcription Translation and Splicing | |
Human, Mouse, Rat, Primate | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction