Learn More
Invitrogen™ HES2 Polyclonal Antibody

Description
HES2 Polyclonal Antibody for IHC (P)
Specifications
Specifications
| Antigen | HES2 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% BSA and 0.05% sodium azide |
| Gene | HES2 |
| Gene Accession No. | O54792, P35429, Q9Y543 |
| Gene Alias | bHLHb40; Class B basic helix-loop-helix protein 40; hairy and enhancer of split 2; hairy and enhancer of split 2 (Drosophila); hes family bHLH transcription factor 2; Hes2; Hes-2; Transcription factor HES-2 |
| Gene Symbols | HES2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.