Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HES2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982193
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2193 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2193 Supplier Bioss Supplier No. BS12383R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Transcription factor HES-2, of the HES family of proteins, contains one basic helix-loop-helix (bHLH) domain and one orange domain. It is reported to express in vascular smooth muscle cells, and may have some role in their differentiation. Activation of Notch signaling pathway leads to activation of Hes family genes through the interaction between Notch intracellular domain and RBPSUH (CSL). Hes family proteins are mammalian homologs of Drosophila Hairy and Enhancer of split. The Hes family of transcriptional repressors are implicated in the cell fate determination of stem cells (or precursor cells) by suppressing the expression of tissue-specific transcriptional activators.

Specifications

Antigen HES2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene HES2
Gene Accession No. Q9Y543
Gene Alias BHLHB40; Class B basic helix-loop-helix protein 40; Hairy and enhancer of split 2; hairy and enhancer of split 2 (Drosophila); hes family bHLH transcription factor 2; HES2; Hes-2; Transcription factor HES-2
Gene Symbols HES2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human HES2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54626
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.