Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HES3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA567801
Description
Predicted to react with Mouse and Rat samples.
Transcriptional repressor of genes that require a bHLH protein for their transcription. Target protein's helix-loop-helix (HLH) protein; may function as repressor in neurons.
Specifications
HES3 | |
Polyclonal | |
Unconjugated | |
HES3 | |
BHLHB43; Class B basic helix-loop-helix protein 43; hairy and enhancer of split 3; hairy and enhancer of split 3 (Drosophila); hes family bHLH transcription factor 3; HES3; Hes-3; Transcription factor HES-3 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
15207, 390992, 64628 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q04667, Q5TGS1, Q61657 | |
HES3 | |
Synthesized peptide derived from human HES3. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction