Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HES3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100761
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100761 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100761 Supplier Invitrogen™ Supplier No. PA5100761
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total HES3.

Transcriptional repressor of genes that require a bHLH protein for their transcription. Target protein's helix-loop-helix (HLH) protein; may function as repressor in neurons.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HES3
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene HES3
Gene Accession No. Q04667, Q5TGS1, Q61657
Gene Alias BHLHB43; Class B basic helix-loop-helix protein 43; hairy and enhancer of split 3; hairy and enhancer of split 3 (Drosophila); hes family bHLH transcription factor 3; HES3; Hes-3; Transcription factor HES-3
Gene Symbols HES3
Host Species Rabbit
Immunogen A synthesized peptide derived from human HES3(Accession Q5TGS1), corresponding to amino acid residues R5-G55.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15207, 390992, 64628
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.