Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HES3 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS12384R

Description
Transcriptional repressor of genes that require a bHLH protein for their transcription. Target protein′s helix-loop-helix (HLH) protein; may function as repressor in neurons.Specifications
HES3 | |
Polyclonal | |
Unconjugated | |
HES3 | |
BHLHB43; Class B basic helix-loop-helix protein 43; Hairy and enhancer of split 3; hairy and enhancer of split 3 (Drosophila); hes family bHLH transcription factor 3; HES3; Hes-3; Transcription factor HES-3 | |
Rabbit | |
Protein A | |
RUO | |
390992 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin) | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q5TGS1 | |
HES3 | |
KLH conjugated synthetic peptide derived from human HES3. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction