Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HES5 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
$196.10 - $499.50
Specifications
Antigen | HES5 |
---|---|
Concentration | 1 mg/mL |
Applications | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
HES5 Polyclonal specifically detects HES5 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
HES5 | |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
Unconjugated | |
RUO | |
PBS with 0.02% Sodium Azide | |
388585 | |
Antibody was raised against a 19 amino acid peptide near the amino terminus of human HES5. The immunogen is located within the first 50 amino acids of HES5. Amino Acid Squence: EKNRLRKPVVEKMRRDRIN | |
Primary | |
At least two isoforms of HES5 are known to exist; this antibody will detect both isoforms. |
1 mg/mL | |
Polyclonal | |
Rabbit | |
Human, Mouse, Rat | |
BHLHB38, bHLHb38Class B basic helix-loop-helix protein 38, Hairy and enhancer of split 5, hairy and enhancer of split 5 (Drosophila), transcription factor HES-5 | |
HES5 | |
IgG | |
Affinity Purified | |
17 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title