Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HES5 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$206.50 - $499.50

Specifications

Antigen HES5
Concentration 1.0 mg/ml
Dilution Western Blot 6-9ug/ml∼, Immunohistochemistry-Paraffin 5ug/ml
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP227174SS
SDP
missing translation for 'viewDocuments'
Novus Biologicals
NBP227174SS
0.025 mg
Each for $206.50
Only null left
Add to Cart
 
NBP227174
SDP
missing translation for 'viewDocuments'
Novus Biologicals
NBP227174
0.1 mg
Each for $499.50
Only null left
Add to Cart
 
Description

Description

HES5 Polyclonal specifically detects HES5 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

HES5
Western Blot 6-9ug/ml∼, Immunohistochemistry-Paraffin 5ug/ml
Polyclonal
Purified
RUO
PBS with 0.05% Sodium Azide
BHLHB38, bHLHb38Class B basic helix-loop-helix protein 38, Hairy and enhancer of split 5, hairy and enhancer of split 5 (Drosophila), transcription factor HES-5
HES5
IgG
Protein A purified
The amino acid sequence used as immunogen is 100% homologous in human, mouse and rat.
1.0 mg/ml
Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Human, Mouse, Rat
Q5TA89
388585
A portion of amino acid1-50 of human HES5 was used as the immunogen for this antibody.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.