Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HES5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA523498
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA523498 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA523498 Supplier Invitrogen™ Supplier No. PA523498
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HES5 Polyclonal Antibody for Western Blot, IHC (P)

Hairy and Enhancer-of-split homologues 5 (HES5) isa helix-loop-helix transcriptional factor that negatively regulates cell differentiation during embryogenesis. It contains one basic helix-loop-helix (bHLH) domain and one Orange domain. HES5 is reported to be a down stream effector of the Notch Signaling Pathway and is expressed in advanced ovarian serous adenocarcinomas as well as in cervical squamous carcinoma. Reports also suggest that HES5 is abundantly expressed in osteoarthritis along with Notch1 and Jagged1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HES5
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene HES5
Gene Accession No. P70120, Q03062, Q5TA89
Gene Alias BHLHB38; Class B basic helix-loop-helix protein 38; hairy and enhancer of split 5; hairy and enhancer of split 5 (Drosophila); hes family bHLH transcription factor 5; Hes5; Hes-5; RP3-395M20.5; Transcription factor HES-5
Gene Symbols HES5
Host Species Rabbit
Immunogen A portion of amino acid1-50 of human HES5.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15208, 388585, 79225
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.