Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HES5 Polyclonal Antibody
GREENER_CHOICE

HES5 Polyclonal Antibody

Supplier:  Thermo Scientific PA550676

Catalog No. PA550676


Only null left
Add to Cart

Description

Description

HES5 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY
Specifications

Specifications

HES5
Polyclonal
Unconjugated
HES5
BHLHB38; Class B basic helix-loop-helix protein 38; hairy and enhancer of split 5; hairy and enhancer of split 5 (Drosophila); hes family bHLH transcription factor 5; Hes5; Hes-5; RP3-395M20.5; Transcription factor HES-5
Rabbit
Antigen affinity chromatography
RUO
15208, 388585, 79225
-20°C
Liquid
Western Blot
1.8 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
P70120, Q03062, Q5TA89
HES5
Synthetic peptide corresponding to a region derived from internal residues of human hes family bHLH transcription factor 5.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.