Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HES6 Antibody (OTI1D6), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272008G
Description
HES6 Monoclonal specifically detects HES6 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
HES6 | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
HES6 | |
Full length recombinant protein of human HES6 (NP_061115) produced in E.coli. | |
0.1 mL | |
55502.0 | |
Store at 4°C in the dark. |
Western Blot | |
OTI1D6 | |
Western Blot | |
BHLHB41, bHLHb41Class B basic helix-loop-helix protein 41, bHLHc23, C-HAIRY1, Hairy and enhancer of split 6, hairy and enhancer of split 6 (Drosophila), HES-6, transcription cofactor HES-6 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction