Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HES7 Antibody, Alexa Fluor™ 647, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP227417AF647
Description
HES7 Polyclonal specifically detects HES7 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
HES7 | |
Polyclonal | |
Western Blot | |
bHLH factor Hes7, BHLHB37, bHLHb37transcription factor HES-7, Class B basic helix-loop-helix protein 37, Hairy and enhancer of split 7, hairy and enhancer of split 7 (Drosophila), hHes7, SCDO4 | |
Rabbit | |
Affinity Purified | |
RUO | |
Human, Mouse, Rat | |
IgG |
Western Blot | |
Alexa Fluor 647 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
HES7 | |
RAENRDGPkMLkPLVEkRc (aa 6-23) | |
0.1 mL | |
Primary | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction