Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HES7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564475
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564475 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564475 Supplier Invitrogen™ Supplier No. PA564475
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PPPPHSQDGA PKAPLPPPPA FWRPWP Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 85%.

HES7 is a member of the hairy and enhancer of split family of bHLH transcription factors. The protein functions as a transcriptional repressor, and is implicated in correct patterning of the axial skeleton.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HES7
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HES7
Gene Accession No. Q9BYE0
Gene Alias bHLH factor Hes7; bHLHb37; class B basic helix-loop-helix protein 37; Hairy and enhancer of split 7; hairy and enhancer of split 7 (Drosophila); hes family bHLH transcription factor 7; HES7; hHes7; mHes7; SCDO4; transcription factor HES-7
Gene Symbols HES7
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human HES7. Recombinant protein control fragment (Product #RP-103993).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84667
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.