Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HES7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA573023
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA573023 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA573023 Supplier Invitrogen™ Supplier No. PA573023
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HES7 Polyclonal Antibody for Western Blot

HES7 is a member of the hairy and enhancer of split family of bHLH transcription factors. The protein functions as a transcriptional repressor, and is implicated in correct patterning of the axial skeleton.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HES7
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene HES7
Gene Accession No. Q8BKT2, Q9BYE0
Gene Alias bHLH factor Hes7; bHLHb37; class B basic helix-loop-helix protein 37; Hairy and enhancer of split 7; hairy and enhancer of split 7 (Drosophila); hes family bHLH transcription factor 7; HES7; hHes7; mHes7; SCDO4; transcription factor HES-7
Gene Symbols HES7
Host Species Rabbit
Immunogen Residues 6-23.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 287423, 84653, 84667
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.