Learn More
Invitrogen™ HESX1 Monoclonal Antibody (2C4)

Description
Peptide Sequence: MSPSLQEGAQ LGENKPSTCS FSIERILGLD QKKDCVPLMK PHRPWADTCS SSGKDGNLCL HVPNPPSGIS FPSVVDHPMP EERASKYENY FSASERLSLK RELSWYRGRR.
Specifications
Specifications
| Antigen | HESX1 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 2C4 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | Hesx1 |
| Gene Accession No. | Q9UBX0 |
| Gene Alias | ANF; Anterior-restricted homeobox protein; CPHD5; HANF; Hes-1; HESX homeobox 1; HESX1; homeo box gene expressed in ES cells; homeobox expressed in ES cells 1; homeobox gene expressed in ES cells; homeobox protein ANF; Rathke pouch homeo box; Rathke pouch homeobox; RGD1563858; Rpx |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.