Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HESX1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA555007
Description
Immunogen sequence: QLGENKPSTC SFSIERILGL DQKKDCVPLM KPHRPWADTC SSSGKDGNLC LHVPNPPSGI SFPSVVDHPM PEERASKYEN YFSASERLSL KRELSWYRGR RPRTAFTQNQ IEVLENVFRV NCYPG Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 79%.
This gene encodes a conserved homeobox protein that is a transcriptional repressor in the developing forebrain and pituitary gland. Mutations in this gene are associated with septooptic dysplasia, HESX1-related growth hormone deficiency, and combined pituitary hormone deficiency.
Specifications
| HESX1 | |
| Polyclonal | |
| Unconjugated | |
| Hesx1 | |
| ANF; Anterior-restricted homeobox protein; CPHD5; HANF; Hes-1; HESX homeobox 1; HESX1; homeo box gene expressed in ES cells; homeobox expressed in ES cells 1; homeobox gene expressed in ES cells; homeobox protein ANF; Rathke pouch homeo box; Rathke pouch homeobox; RGD1563858; Rpx | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 8820 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9UBX0 | |
| Hesx1 | |
| Recombinant protein corresponding to Human HESX1. Recombinant protein control fragment (Product #RP-89385). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction