Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HESX1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983010
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3010 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3010 Supplier Bioss Supplier No. BS13597R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a conserved homeobox protein that is a transcriptional repressor in the developing forebrain and pituitary gland. Mutations in this gene are associated with septooptic dysplasia, HESX1-related growth hormone deficiency, and combined pituitary hormone deficiency.

Specifications

Antigen HESX1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Hesx1
Gene Accession No. Q9UBX0
Gene Alias ANF; Anterior-restricted homeobox protein; CPHD5; HANF; Hes-1; HESX homeobox 1; Hesx1; homeo box gene expressed in ES cells; Homeobox expressed in ES cells 1; homeobox gene expressed in ES cells; Homeobox protein ANF; Rathke pouch homeo box; Rathke pouch homeobox; RGD1563858; Rpx
Gene Symbols Hesx1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human HESX1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8820
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.