Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heterochromatin Protein-1 Mouse anti-Human, Mouse, MilliporeSigma™

Catalog No. MABE66625UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE666-25UG 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE666-25UG Supplier MilliporeSigma Supplier No. MABE66625UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Heterochromatin Protein-1 , clone 4C12.1 is a highly specific mouse monoclonal antibody, that targets Heterochromatin Protein-1 alpha and has been tested in Western Blotting and Immunohistochemistry (Paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Heterochromatin Protein-1
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols CBX5,HP1A
Host Species Mouse
Immunogen GST/His-tagged recombinant fraction corresponding to 68 amino acids from the internal region of human Heterochromatin protein 1 homolog alpha.
Purification Method Protein G Purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.