Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heterogeneous Nuclear Ribonucleoprotein (A1-like) Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NBP180446

Catalog No. NBP180446


Only null left
Add to Cart

Description

Description

Heterogeneous Nuclear Ribonucleoprotein (A1-like) Polyclonal specifically detects Heterogeneous Nuclear Ribonucleoprotein (A1-like) in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

Heterogeneous Nuclear Ribonucleoprotein (A1-like)
Polyclonal
Unconjugated
PBS, 2% Sucrose with 0.09% Sodium Azide
heterogeneous nuclear ribonucleoprotein A1-like 2, hnRNP A1-like 2, hnRNP core protein A1-like 2, HNRNPA1L, LOC144983, MGC102957
Rabbit
Affinity purified
RUO
144983
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
0.5 mg/ml
Western Blot 1.0 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
NP_001011724
HNRNPA1L2
Synthetic peptide directed towards the N terminal of human RP11-78J21. 1. Peptide sequence MSKSASPKEPEQLRKLFIGGLSFETTDESLRSHFEQWGTLTDCVVMRDPN.
100 μL
Primary
Expected identity based on immunogen sequence: Chimpanzee: 100%; Equine: 100%; Chicken: 100%; Crab-eating macaque: 100%; Rhesus macaque: 100%; Pig: 100%; Mouse: 100%; Canine: 100%; Bovine: 100%; Rat: 100%; Zebrafish: 92%; Western clawed frog: 92%; Green puffer: 92%; Japanese pufferfish: 92%; Xenopus: 92%; Atlantic salmon: 92%; Burton's mouthbrooder: 92%; Greater Equineshoe bat: 85%; Human: 100%; Sumatran orangutan: 84%; American grasshopper: 83%; Body louse: 81%; Rainbow trout: 76%; Fission yeast: 76%.
Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only