Learn More
Hex Mouse anti-Human, Clone: OTI2H7 2c2, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF500098

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
HHEX is a member of the homeobox family of transcription factors, many of which are involved in developmental processes. Expression in specific hematopoietic lineages suggests that this protein may play a role in hematopoietic differentiation. Establishes anterior identity at two levels 1) acts early to enhance canonical WNT-signaling by repressing expression of TLE4 and 2) acts later to inhibit NODAL-signaling by directly targeting NODAL.Specifications
Hex | |
Monoclonal | |
Unconjugated | |
HHEX | |
HEX, HMPH, HOX11L-PEN, PRH, PRHX | |
Mouse | |
Affinity Chromatography | |
RUO | |
3087 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI2H7 2c2 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q03014 | |
HHEX | |
Recombit fragment expressed in E.coli corresponding to amino acids 100-270 of human Hex | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.