Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hex Mouse anti-Human, Clone: OTI3E6, liquid, TrueMAB™
SDP

Catalog No. 501671433
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-1433 30 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-167-1433 Supplier OriGene Supplier No. TA500219S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HHEX is a member of the homeobox family of transcription factors, many of which are involved in developmental processes. Expression in specific hematopoietic lineages suggests that this protein may play a role in hematopoietic differentiation. Establishes anterior identity at two levels 1) acts early to enhance canonical WNT-signaling by repressing expression of TLE4 and 2) acts later to inhibit NODAL-signaling by directly targeting NODAL.

Specifications

Antigen Hex
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone OTI3E6
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene HHEX
Gene Accession No. Q03014
Gene Alias HEX, HMPH, HOX11L-PEN, PRH, PRHX
Gene Symbols HHEX
Host Species Mouse
Immunogen Recombit fragment expressed in E.coli corresponding to amino acids 100-270 of human Hex
Purification Method Affinity Chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3087
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.