Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Hex Recombinant Rabbit Monoclonal Antibody (9P4E4)
GREENER_CHOICE

Catalog No. PIMA537951
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA537951 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA537951 Supplier Invitrogen™ Supplier No. MA537951
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive Samples: HepG2 Immunogen sequence: TLPSPNSSFT SLVSPYRTPV YEPTPIHPAF SHHSAAALAA AYGPGGFGGP LYPFPRTVND YTHALLRHDP LGKPLLWSPF LQRPLHKRKG GQVRFSNDQT I.

HHEX is a member of the homeobox family of transcription factors, many of which are involved in developmental processes. Expression in specific hematopoietic lineages suggests that this protein may play a role in hematopoietic differentiation. Establishes anterior identity at two levels 1) acts early to enhance canonical WNT-signaling by repressing expression of TLE4 and 2) acts later to inhibit NODAL-signaling by directly targeting NODAL.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Hex
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 9P4E4
Concentration 0.46 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene HHEX
Gene Accession No. Q03014
Gene Alias hematopoietically expressed homeobox; hematopoietically expressed homeobox, related sequence 2; hematopoietically-expressed homeobox protein Hhex; HEX; Hex1; HHEX; Hhex-rs2; HMPH; homeobox; homeobox protein HEX; Homeobox protein PRH; homeobox, hematopoietically expressed; HOX11L-PEN; mHex; Prh; Prhx; proline-rich homeodomain-containing transcription factor
Gene Symbols HHEX
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 50-150 of human Hex (Q03014).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3087
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.