Alfa Aesar™ Hexa-n-butylditin, 98%

Manufacturer: Alfa Aesar™ A1200709

Quantity 10g

 View more versions of this product

Catalog No. AAA1200709

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 813-19-4
    Molecular Formula C24H54Sn2
    Linear Formula (CH3CH2CH2CH2)3Sn-Sn(CH2CH2CH2CH3)3
    Formula Weight 580.07
    MDL Number MFCD00009417
    UN Number UN2788
    Beilstein 3605476
    Synonym Bis(tri-n-butyltin); Hexa-n-butyldistannane
    Chemical Name or Material Hexa-n-butylditin

    Specifications

    Formula Weight 580.07
    Health Hazard 3 GHS P Statement
    P260-P280-P301+P310-P305+P351+P338-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula C24H54Sn2
    UN Number UN2788
    MDL Number MFCD00009417
    Beilstein 3605476
    Synonym Bis(tri-n-butyltin); Hexa-n-butyldistannane
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.
    Recommended Storage −30°C to −10°C
    Linear Formula (CH3CH2CH2CH2)3Sn-Sn(CH2CH2CH2CH3)3
    CAS 813-19-4
    Quantity 10g
    Flash Point 124°C (255°F)
    Boiling Point 197° to 198°C (10mmHg)
    Health Hazard 2 GHS H Statement
    H301-H372-H312-H315-H319
    Toxic if swallowed.
    Causes damage to organs through prolonged or repeated exposure.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    Chemical Name or Material Hexa-n-butylditin
    Assay 98%

    Description

    Applications
    Hexa-n-butylditin is used in palladium-catalyzed tin-carbon bond formation. It finds useful application in deoxygenation and desulfurization reactions. It acts as a source of tributylstannyl radicals in synthetic chemistry. It is also used in the radioiodination of styrlpyridines. Further, it is used as a reagent to stannylate aryl halides for further Stille coupling.

    Solubility
    Immiscible with water.

    Notes
    Incompatible with strong oxidizing agents.

    GHS H Statement
    H301-H372-H312-H315-H319
    Toxic if swallowed.
    Causes damage to organs through prolonged or repeated exposure.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.

    −30°C to −10°C

    GHS P Statement
    P260-P280-P301+P310-P305+P351+P338-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit