Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HEXA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA579357
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA579357 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA579357 Supplier Invitrogen™ Supplier No. PA579357
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: Rat Liver Tissue, HELA whole cell, SMMC whole cell. IHC: Human Intestinal Cancer Tissue.

This gene encodes the alpha subunit of the lysosomal enzyme beta-hexosaminidase that, together with the cofactor GM2 activator protein, catalyzes the degradation of the ganglioside GM2, and other molecules containing terminal N-acetyl hexosamines. Beta-hexosaminidase is composed of two subunits, alpha and beta, which are encoded by separate genes. Both beta-hexosaminidase alpha and beta subunits are members of family 20 of glycosyl hydrolases. Mutations in the alpha or beta subunit genes lead to an accumulation of GM2 ganglioside in neurons and neurodegenerative disorders termed the GM2 gangliosidoses. Alpha subunit gene mutations lead to Tay-Sachs disease (GM2-gangliosidosis type I).
TRUSTED_SUSTAINABILITY

Specifications

Antigen HEXA
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 5mg BSA and 0.05mg sodium azide, 0.05mg thimerosal
Gene HEXA
Gene Accession No. P06865, P29416, Q641X3
Gene Alias beta-hexosaminidase alpha-subunit; beta-hexosaminidase subunit alpha; Beta-hexosaminidase subunit beta; Beta-hexosaminidase subunit beta chain A; Beta-hexosaminidase subunit beta chain B; Beta-N-acetylhexosaminidase subunit alpha; beta-N-acetylhexosaminidase subunit beta; Cervical cancer proto-oncogene 7 protein; EC 3.2.1; EC 3.2.1.52; ENC-1AS; epididymis luminal protein 248; epididymis secretory protein Li 111; glycosidase; HCC7; HCC-7; HEL-248; HEL-S-111; Hex-1; HEXA; Hexb; hexosaminidase A; hexosaminidase A (alpha polypeptide); hexosaminidase A alpha; hexosaminidase B; hexosaminidase B (beta polypeptide); hexosaminidase subunit A; hexosaminidase subunit alpha; hexosaminidase subunit B; hexosaminidase subunit beta; Hexose aminidase A (alpha polypeptide); MGC99608; N-acetyl-beta-glucosaminidase subunit alpha; N-acetyl-beta-glucosaminidase subunit beta; TSD
Gene Symbols HEXA
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence in the middle region of human HEXA (191-207aa DVMAYNKLNVFHWHLVD).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15211, 300757, 3073
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.