Learn More
Invitrogen™ HEXA Polyclonal Antibody

Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human Hela whole cell, human 293T whole cell, human Caco-2 whole cell, human THP-1 whole cell, human HepG2 whole cell.
Specifications
Specifications
| Antigen | HEXA |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 500 μg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5mg BSA and 0.05mg sodium azide, 0.05mg thimerosal |
| Gene | HEXA |
| Gene Accession No. | P06865 |
| Gene Alias | beta-hexosaminidase alpha-subunit; beta-hexosaminidase subunit alpha; Beta-hexosaminidase subunit beta; Beta-hexosaminidase subunit beta chain A; Beta-hexosaminidase subunit beta chain B; Beta-N-acetylhexosaminidase subunit alpha; beta-N-acetylhexosaminidase subunit beta; Cervical cancer proto-oncogene 7 protein; EC 3.2.1; EC 3.2.1.52; ENC-1AS; epididymis luminal protein 248; epididymis secretory protein Li 111; glycosidase; HCC7; HCC-7; HEL-248; HEL-S-111; Hex-1; HEXA; Hexb; hexosaminidase A; hexosaminidase A (alpha polypeptide); hexosaminidase A alpha; hexosaminidase B; hexosaminidase B (beta polypeptide); hexosaminidase subunit A; hexosaminidase subunit alpha; hexosaminidase subunit B; hexosaminidase subunit beta; Hexose aminidase A (alpha polypeptide); MGC99608; N-acetyl-beta-glucosaminidase subunit alpha; N-acetyl-beta-glucosaminidase subunit beta; TSD |
| Gene Symbols | HEXA |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.