Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HEXA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5139763
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5139763 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5139763 Supplier Invitrogen™ Supplier No. PA5139763
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: LESFYIQTLL DIVSSYGKGY VVWQEVFDNK VKIQPDTIIQ VWREDIPVNY MKELELVTKA GFRALLSAPW YLNRISYGPD WKDFYVVEPL AFE Highest antigen sequence indentity to the following orthologs: Mouse - 81%, Rat - 82%.

This gene encodes the alpha subunit of the lysosomal enzyme beta-hexosaminidase that, together with the cofactor GM2 activator protein, catalyzes the degradation of the ganglioside GM2, and other molecules containing terminal N-acetyl hexosamines. Beta-hexosaminidase is composed of two subunits, alpha and beta, which are encoded by separate genes. Both beta-hexosaminidase alpha and beta subunits are members of family 20 of glycosyl hydrolases. Mutations in the alpha or beta subunit genes lead to an accumulation of GM2 ganglioside in neurons and neurodegenerative disorders termed the GM2 gangliosidoses. Alpha subunit gene mutations lead to Tay-Sachs disease (GM2-gangliosidosis type I).
TRUSTED_SUSTAINABILITY

Specifications

Antigen HEXA
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HEXA
Gene Accession No. P06865
Gene Alias beta-hexosaminidase alpha-subunit; beta-hexosaminidase subunit alpha; Beta-hexosaminidase subunit beta; Beta-hexosaminidase subunit beta chain A; Beta-hexosaminidase subunit beta chain B; Beta-N-acetylhexosaminidase subunit alpha; beta-N-acetylhexosaminidase subunit beta; Cervical cancer proto-oncogene 7 protein; EC 3.2.1; EC 3.2.1.52; ENC-1AS; epididymis luminal protein 248; epididymis secretory protein Li 111; glycosidase; HCC7; HCC-7; HEL-248; HEL-S-111; Hex-1; HEXA; Hexb; hexosaminidase A; hexosaminidase A (alpha polypeptide); hexosaminidase A alpha; hexosaminidase B; hexosaminidase B (beta polypeptide); hexosaminidase subunit A; hexosaminidase subunit alpha; hexosaminidase subunit B; hexosaminidase subunit beta; Hexose aminidase A (alpha polypeptide); MGC99608; N-acetyl-beta-glucosaminidase subunit alpha; N-acetyl-beta-glucosaminidase subunit beta; TSD
Gene Symbols HEXA
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human HEXA. Recombinant protein control fragment (Product #RP-91303).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3073
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.