Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexaamminecobalt(III) chloride, 99.999%, (trace metal basis), ACROS Organics™

Click to view available options
Quantity:
5g
10g
Packaging:
Glass bottle
10534-89-1
H18Cl3CoN6
267.47
MFCD00036304
10534-89-1
H18Cl3CoN6
267.47
MFCD00036304
for ultra trace analysis
Melting Point 217°C
Total Trace Metal Impurities 0.001% max
Color Orange to Brown
Quantity 5g
Packaging Glass bottle
Assay Percent Range 99.999%
Linear Formula [Co(NH3)6]Cl3
Merck Index 15, 4710
Solubility Information Solubility in water: soluble
Formula Weight 267.47
Physical Form Powder
Percent Purity 100.00%
Grade Trace Metal Basis
Chemical Name or Material Hexaamminecobalt(III) chloride, 99.999%
Show More Show Less

GHS H Statement
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of causing cancer if inhaled.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
In case of inadequate ventilation wear respiratory protection.
IF INHALED: If breathing is di

GHS Signal Word: Danger

EINECSNumber : 234-103-9

TSCA : TSCA