Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexacarbonylmolybdenum, 98%

Catalog No. p-7049150
Click to view available options
Quantity:
2.5 g
25 g
100 g

CAS: 13939-06-5 | C6MoO6 | 264.01 g/mol

Hexacarbonylmolybdenum is widely utilized in electron beam-induced deposition technique providing an appropriate source of molybdenum atoms. It finds an application as important reagent in organometallic synthesis because of replacement of carbonyl ligands by other donor ligands. Its derivative is act as a catalyst in organic synthesis, which is used in alkyne metathesis and the Pauson-Khand reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13939-06-5
Molecular Formula C6MoO6
Molecular Weight (g/mol) 264.01
MDL Number MFCD00003466
InChI Key KMKBZNSIJQWHJA-UHFFFAOYSA-N
Synonym molybdenumhexacarbonyl, carbon monooxide-molybdenum 6:1, hexakis carbon monoxide molybdenum, hexacarbonylmolybdenu, hexakis methanidylidyneoxidanium molybdenum, hexacarbonylmolybdenum, may contain up to cr 5g
PubChem CID 2724068
ChEBI CHEBI:30508
IUPAC Name carbon monoxide;molybdenum
SMILES [C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[Mo]

Specifications

Melting Point ∽150°C (decomposition)
Physical Form Crystalline
Quantity 2.5 g
Assay Percent Range 98%
Linear Formula Mo(CO)6
UN Number UN3466
Solubility Information Slightly soluble in tetrahydrofuran,diglyme and acetonitrile.
Formula Weight 264
Percent Purity 98%
Density 1.96
Chemical Name or Material Hexacarbonylmolybdenum, Up to 1% Cr
Show More Show Less
Hazard Category H300+H310+H330
Hazard Statement GHS H Statement
H300-H330
Precautionary Statement P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P310-P330-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: METAL CARBONYLS, SOLID, N.O.S.
EINECSNumber 237-713-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.