Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexadecafluorosebacic Acid 96.0+%, TCI America™
SDP

Catalog No. H089225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H089225G 25 g
H08925G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H089225G Supplier TCI America Supplier No. H089225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 307-78-8
Molecular Formula C10H2F16O4
Molecular Weight (g/mol) 490.096
MDL Number MFCD00042365
InChI Key YPCSMEGZIYWAAZ-UHFFFAOYSA-N
Synonym Hexadecafluorodecanedioic Acid, Perfluorodecanedioic Acid, Perfluorosebacic Acid
PubChem CID 2733269
IUPAC Name 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9-hexadecafluorodecanedioic acid
SMILES C(=O)(C(C(C(C(C(C(C(C(C(=O)O)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)O

Specifications

Melting Point 162°C
Color White-Yellow
Quantity 25 g
Formula Weight 490.10
Percent Purity ≥96.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Hexadecafluorosebacic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.