Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hexadecane, Reagent, 99%, Spectrum™ Chemical

Click to view available options
Quantity:
100 mL
500 mL
18.1 L
200 L
Packaging:
Amber Glass Bottle
Drum
Steel Pail
Description
Spectrum™ Chemical Hexadecane, Reagent
Specifications
Specifications
CAS Max % | 100% |
Packaging | Amber Glass Bottle |
Quantity | 500 mL |
Percent Purity | 99% |
Grade | Reagent |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction