Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexadecyl Gallate 95.0+%, TCI America™
SDP

Catalog No. G001325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G001325G 25 g
G00135G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G001325G Supplier TCI America Supplier No. G001325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5026-65-3
Molecular Formula C23H38O5
Molecular Weight (g/mol) 394.55
MDL Number MFCD00016428
InChI Key TYCUSKFOGZNIBO-UHFFFAOYSA-N
Synonym Cetyl Gallate, Gallic Acid Cetyl Ester, Gallic Acid Hexadecyl Ester
PubChem CID 78729
IUPAC Name hexadecyl 3,4,5-trihydroxybenzoate
SMILES CCCCCCCCCCCCCCCCOC(=O)C1=CC(O)=C(O)C(O)=C1

Specifications

Melting Point 100°C
Quantity 25 g
Formula Weight 394.55
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Hexadecyl Gallate
EINECSNumber (3)-1584
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.