Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexadecylcyclohexane 97.0+%, TCI America™
SDP

Catalog No. H096625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H096625G 25 g
H09665G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H096625G Supplier TCI America Supplier No. H096625G

Chemical Identifiers

CAS 6812-38-0
Molecular Formula C22H44
Molecular Weight (g/mol) 308.59
MDL Number MFCD04038421
InChI Key NRHBFNBZAZZTAU-UHFFFAOYSA-N
Synonym 1-Cyclohexylhexadecane
PubChem CID 23250
IUPAC Name hexadecylcyclohexane
SMILES CCCCCCCCCCCCCCCCC1CCCCC1

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 308.59
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Hexadecylcyclohexane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.