Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexadecyltrimethylammonium Bromide (Technical), Fisher Chemical

Catalog No. o3042500 Shop All Fisherbrand Products
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle

Chemical Identifiers

CAS 57-09-0
Molecular Formula C19H42BrN
Molecular Weight (g/mol) 364.46
MDL Number MFCD00011772
InChI Key LZZYPRNAOMGNLH-UHFFFAOYSA-M
Synonym cetrimonium bromide, cetyltrimethylammonium bromide, hexadecyltrimethylammonium bromide, ctab, centimide, cetab, bromat, ctmab, cee dee, cetavlon
PubChem CID 5974
ChEBI CHEBI:3567
IUPAC Name hexadecyltrimethylazanium bromide
SMILES [Br-].CCCCCCCCCCCCCCCC[N+](C)(C)C

Specifications

Decomposition Information >230°C
Melting Point 230°C
Color White
pH 5 to 7
Flash Point 244°C
Packaging Poly Bottle
Linear Formula CH3(CH2)15N(CH3)3Br
Quantity 500 g
UN Number 3261
Vapor Pressure Negligible
Formula Weight 364.46
Grade Technical
Physical Form Powder or Solid
Chemical Name or Material Hexadecyltrimethylammonium Bromide
Show More Show Less
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Harmful if swallowed. Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Hygroscopic. Use personal protective equipment. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Obtain medical attention. Obtain medical attention. Do not induce vomiting.

NFPA

Health:2
Flammability:1
Instability:1
DOTInformation Class 8: Corrosive
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.