Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexadecyltrimethylammonium Chloride, 97%

Catalog No. ac411410050
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass Bottle
Plastic bottle

CAS: 112-02-7 | C19H42ClN | 320.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-02-7
Molecular Formula C19H42ClN
Molecular Weight (g/mol) 320.00
MDL Number MFCD00011773
InChI Key WOWHHFRSBJGXCM-UHFFFAOYSA-M
Synonym hexadecyltrimethylammonium chloride, cetrimonium chloride, cetyltrimethylammonium chloride, n,n,n-trimethylhexadecan-1-aminium chloride, dehyquart a, cetyl trimethyl ammonium chloride, aliquat 6, trimethylcetylammonium chloride, arquad 16-50, n-hexadecyltrimethylammonium chloride
PubChem CID 8154
ChEBI CHEBI:53581
IUPAC Name hexadecyl(trimethyl)azanium;chloride
SMILES [Cl-].CCCCCCCCCCCCCCCC[N+](C)(C)C

Specifications

Melting Point 234°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (Argentometry)
Packaging Glass Bottle
Linear Formula CH3(CH2)15N(Cl)(CH3)3
Quantity 5 g
Solubility Information Solubility in water: soluble
Formula Weight 319.75
Percent Purity 97%
Physical Form Amorphous Powder
Chemical Name or Material Hexadecyltrimethylammonium chloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF SWALLOWED
EINECSNumber 203-928-6

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.