Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexaethylphosphorus Triamide, 95%

Catalog No. AC423080050
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC423080050 5 g Glass bottle
AC423080250 25 g Glass bottle
AC423081000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. AC423080050 Supplier Thermo Scientific Chemicals Supplier No. 423080050
Add to Cart
This item is not returnable. View return policy

CAS: 2283-11-6 | C12H30N3P | 247.37 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2283-11-6
Molecular Formula C12H30N3P
Molecular Weight (g/mol) 247.37
MDL Number MFCD00009041
InChI Key FDIOSTIIZGWENY-UHFFFAOYSA-N
Synonym tris diethylamino phosphine, hexaethylphosphorous triamide, hexaethyl phosphorous triamide, phosphorous triamide, hexaethyl, n,n,n',n',n,n-hexaethylphosphinetriamine, tris n,n-diethylamino phosphine, hexaethyltriaminophosphine, hexaethylphosphoruo triamide, hexaethylphosphorus triamide, n-bis diethylamino phosphanyl-n-ethylethanamine
PubChem CID 75292
IUPAC Name [bis(diethylamino)phosphanyl]diethylamine
SMILES CCN(CC)P(N(CC)CC)N(CC)CC

Specifications

Color Colorless to Orange-Yellow
Density 0.8900g/mL
Boiling Point 111°C (6.0 mmHg)
Flash Point 58°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula ((C2H5)2N)3P
Refractive Index 1.4720 to 1.475
Quantity 5 g
Specific Gravity 0.89
Solubility Information Solubility in water: insoluble
Formula Weight 247.36
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Hexaethylphosphorus triamide
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wash face,hands and any exposed skin thoroughly
EINECSNumber 218-920-8

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.