Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexafluoroacetone Deuterate 95.0atom%D, TCI America™
SDP

Catalog No. H06935G
Click to view available options
Quantity:
5 g

Chemical Identifiers

CAS 109640-39-3
Molecular Formula C15H16F30O13
Molecular Weight (g/mol) 974.23
MDL Number MFCD09953659,MFCD09953659,MFCD00149064
InChI Key MXXAOJTWEBEIPQ-UHFFFAOYSA-N
Synonym hexafluoroacetone deuterate 95.0atom%d
PubChem CID 131844136
IUPAC Name pentakis(hexafluoropropan-2-one) octahydrate
SMILES O.O.O.O.O.O.O.O.FC(F)(F)C(=O)C(F)(F)F.FC(F)(F)C(=O)C(F)(F)F.FC(F)(F)C(=O)C(F)(F)F.FC(F)(F)C(=O)C(F)(F)F.FC(F)(F)C(=O)C(F)(F)F

Specifications

Color Colorless
UN Number 2552
Quantity 5 g
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material Hexafluoroacetone Deuterate 95.0atom%D
EINECSNumber (2)-0581
RTECSNumber UC2700000
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.