Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

gamma-Hexalactone, 98%

Catalog No. p-7077259
Click to view available options
Quantity:
25 g
100 g
500 g

CAS: 695-06-7 | C6H10O2 | 114.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 695-06-7
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.14
MDL Number MFCD00005401
InChI Key JBFHTYHTHYHCDJ-UHFFFAOYNA-N
Synonym gamma-caprolactone, gamma-hexalactone, 4-hexanolide, gamma-hexanolactone, 6-caprolactone, tonkalide, toukalide, 2 3h-furanone, 5-ethyldihydro, 4-ethyl-4-butanolide, 5-ethyldihydrofuran-2 3h-one
PubChem CID 12756
ChEBI CHEBI:85235
IUPAC Name 5-ethyloxolan-2-one
SMILES CCC1CCC(=O)O1

Specifications

Melting Point -18.0°C
Density 1.0230g/mL
Boiling Point 220.0°C
Flash Point 98°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Quantity 25 g
Specific Gravity 1.023
Solubility Information Solubility in water: slightly soluble.
Formula Weight 114.14
Percent Purity 98%
Chemical Name or Material γ-Hexalactone
Show More Show Less
EINECSNumber 211-778-8

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.