Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexamethonium Chloride Dihydrate 99.0+%, TCI America™
SDP

Catalog No. H0090100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0090100G 100 g
H009025G 25 g
H0090500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H0090100G Supplier TCI America Supplier No. H0090100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 60-25-3
Molecular Formula C12H32N2O2
Molecular Weight (g/mol) 236.40
MDL Number MFCD00031563
InChI Key GYLUMIIRFKDCKI-UHFFFAOYSA-L
Synonym hexamethonium chloride, hexamethonium dichloride, chloor-hexaviet, esomid chloride, hexone chloride, hiohex chloride, meton, hexon chloride, methium chloride, bistrium chloride
PubChem CID 93550
IUPAC Name trimethyl[6-(trimethylazaniumyl)hexyl]azanium dihydroxide
SMILES [OH-].[OH-].C[N+](C)(C)CCCCCC[N+](C)(C)C

Specifications

Melting Point 292°C
Color White
Quantity 100 g
Formula Weight 273.29
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Hexamethonium Chloride Dihydrate
EINECSNumber (2)-0180
RTECSNumber BQ8650000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.