Learn More
Hexamethylcyclotrisiloxane, 97%
Description
Hexamethylcyclotrisiloxane is used in the preparation of graft polymers and block polymers. It is also a useful biochemical for proteomics research and also acts as an intermediate in organic reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Hexamethylcyclotrisiloxane is used in the preparation of graft polymers and block polymers. It is also a useful biochemical for proteomics research and also acts as an intermediate in organic reactions.
Solubility
Slightly soluble in water.
Notes
Moisture sensitive. Incompatible in strong oxidizing agents.

Specifications
Specifications
Boiling Point | 134°C |
Melting Point | 64°C to 66°C |
Quantity | 100 g |
UN Number | UN1325 |
Beilstein | 1766829 |
Flash Point | 35°C (95°F) |
Solubility Information | Slightly soluble in water. |
Formula Weight | 222.47 |
Percent Purity | 97% |
Sensitivity | Moisture sensitive |
Show More |
Safety and Handling
Hazard Category | H228 |
Hazard Statement | GHS H Statement H228-H315-H319-H335 Flammable solid. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P210-P240-P241-P280-P370+P378q |
DOTInformation | Hazard Class: 4.1; Packaging Group: III |
EINECSNumber | 208-765-4 |
RTECSNumber | GZ4705000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.