Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexamethylcyclotrisiloxane, 98%

Catalog No. p-141998
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 541-05-9 | C6H18O3Si3 | 222.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 541-05-9
Molecular Formula C6H18O3Si3
Molecular Weight (g/mol) 222.46
MDL Number MFCD00005943
InChI Key HTDJPCNNEPUOOQ-UHFFFAOYSA-N
Synonym hexamethylcyclotrisiloxane, cyclotrisiloxane, hexamethyl, dimethylsiloxane cyclic trimer, unii-epv75l8o0r, ccris 1326, sdk 10, hexamethylcyclohexasiloxane, epv75l8o0r, cyclotrisiloxane, 2,2,4,4,6,6-hexamethyl, dsstox_cid_7185
PubChem CID 10914
SMILES C[Si]1(C)O[Si](C)(C)O[Si](C)(C)O1

Specifications

Melting Point 63.0°C to 67.0°C
Color Colorless to White
Boiling Point 134.0°C
Flash Point 35°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 25 g
Beilstein 04,III,1884
Merck Index 09,4559
IUPAC Name 2,2,4,4,6,6-hexamethyl-1,3,5,2,4,6-trioxatrisilinane
Formula Weight 222.47
Percent Purity 98%
Physical Form Crystalline Chunks
Chemical Name or Material Hexamethylcyclotrisiloxane, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Flammable solid.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Store in a well-ventilated place.
Keep container tightly closed.
EINECSNumber 208-765-4
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.