Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexamethyldisilazane, 98+%

Catalog No. p-7022387
Click to view available options
Quantity:
25 mL
100 mL
500 mL
2500 mL

CAS: 999-97-3 | C6H19NSi2 | 161.395 g/mol

Hexamethyldisilazane is used as a solvent in organic synthesis and organometallic chemistry. It is often used as an adhesion promoter for photoresist in photolithography. Further, it is used for the preparation of trimethylsilyl ethers from hydroxy compounds. It is used as an alternative to critical point drying during sample preparation in electron microscopy. It is added to analyte to get silylated diagnostic products during pyrolysis in gas chromatography- mass spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hexamethyldisilazane is used as a solvent in organic synthesis and organometallic chemistry. It is often used as an adhesion promoter for photoresist in photolithography. Further, it is used for the preparation of trimethylsilyl ethers from hydroxy compounds. It is used as an alternative to critical point drying during sample preparation in electron microscopy. It is added to analyte to get silylated diagnostic products during pyrolysis in gas chromatography- mass spectrometry.

Solubility
Miscible with methanol, chloroform, ethyl ether and benzene. Immiscible with water.                    

Notes
Incompatible with strong oxidizing agents and strong acids.  
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 999-97-3
Molecular Formula C6H19NSi2
Molecular Weight (g/mol) 161.395
MDL Number MFCD00008259
InChI Key FFUAGWLWBBFQJT-UHFFFAOYSA-N
Synonym hexamethyldisilazane, bis trimethylsilyl amine, hmds, 1,1,1,3,3,3-hexamethyldisilazane, hexamethylsilazane, silanamine, 1,1,1-trimethyl-n-trimethylsilyl, tri-sil, 1,1,1-trimethyl-n-trimethylsilyl silanamine, hexamethyldisilizane, disilazane, 1,1,1,3,3,3-hexamethyl
PubChem CID 13838
ChEBI CHEBI:85068
IUPAC Name [dimethyl-(trimethylsilylamino)silyl]methane
SMILES C[Si](C)(C)N[Si](C)(C)C

Specifications

Melting Point -78°C
Density 0.774
Boiling Point 126°C
Flash Point 8°C (46°F)
Odor Odorless
Assay Percent Range 98+%
Refractive Index 1.408
Linear Formula (CH3)3SiNHSi(CH3)3
Quantity 25 mL
UN Number UN3286
Beilstein 635752
Sensitivity Moisture sensitive
Merck Index 14,4689
Solubility Information Miscible with methanol,chloroform,ethyl ether and benzene. Immiscible with water.
Formula Weight 161.4
Percent Purity ≥98%
Physical Form Liquid
Chemical Name or Material Hexamethyldisilazane
Show More Show Less
Hazard Category H225-H302+H332-H311-H314-H335
Hazard Statement GHS H Statement
H225-H311-H331-H314-H318-H302
Highly flammable liquid and vapour.
Toxic in contact with skin.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUID, TOXIC, CORROSIVE, N.O.S.
EINECSNumber 213-668-5
RTECSNumber JM9230000
TSCA Yes
storageNote1 Moisture sensitive
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.