Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexamethyldisiloxane, 98+%

Catalog No. p-7081743
Click to view available options
Quantity:
100 mL
500 mL
2500 mL

CAS: 107-46-0 | C6H18OSi2 | 162.379 g/mol

Hexamethyldisiloxane is used as a solvent in organic reactions. It finds application as an international standard for 1H NMR spectroscopy for calibrating chemical shift. Further, it is used in liquid bandages to protect skin from irritation. In addition, it is used to measure tissue oxygen tension as a receptor molecule. It is also used to make low-k dielectric materials for the semi-conductor industries by plasma-enhanced chemical vapor deposition(PECVD) method.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hexamethyldisiloxane is used as a solvent in organic reactions. It finds application as an international standard for 1H NMR spectroscopy for calibrating chemical shift. Further, it is used in liquid bandages to protect skin from irritation. In addition, it is used to measure tissue oxygen tension as a receptor molecule. It is also used to make low-k dielectric materials for the semi-conductor industries by plasma-enhanced chemical vapor deposition(PECVD) method.

Solubility
Slightly miscible with water.

Notes
Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, acids, bases and oxygen.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-46-0
Molecular Formula C6H18OSi2
MDL Number MFCD00008265
InChI Key UQEAIHBTYFGYIE-UHFFFAOYSA-N
Synonym hexamethyldisiloxane, disiloxane, hexamethyl, hexamethyl disiloxane, oxybis trimethylsilane, fluka ag, hmdso, bis trimethylsilyl ether, belsil dm 0.65, bis trimethylsilyl oxide
PubChem CID 24764
ChEBI CHEBI:78002
IUPAC Name trimethyl(trimethylsilyloxy)silane
SMILES C[Si](C)(C)O[Si](C)(C)C

Specifications

Melting Point -67°C
Density 0.764
Boiling Point 100°C to 101°C
Flash Point −2°C (28°F)
Odor Characteristic
Refractive Index 1.377
Linear Formula (CH3)3SiOSi(CH3)3
Quantity 100 mL
UN Number UN1993
Beilstein 1736258
Solubility Information Slightly miscible with water.
Molecular Weight (g/mol) 162.379
Formula Weight 162.38
Percent Purity ≥98%
Chemical Name or Material Hexamethyldisiloxane
Show More Show Less
Hazard Category H225
Hazard Statement GHS H Statement
H225
Highly flammable liquid and vapor.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P271-P280-P303+P361+P353-P304+P340-P312-P314-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 203-492-7
RTECSNumber JM9237000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.